HSP70B' ELISA kit
Supplier: ENZO LIFE SCIENCES
ADIEKS725A
ENZOADIEKS725AEA 1090 EUR
ENZOADIEKS725A
HSP70B' ELISA kit
Assays ELISAs
The HSP70B' EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 4.5 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.Only commercially available ELISA kit for ultra-sensitive HSP70B' detection in cell lysates, serum, and tissue.
  • Only source for HSP70B’ ELISA, due to patent protection
  • Precious sample compared to Western blot analysis
  • Fast and quantitative results
For the quantitative determination of HSP70B' in cell lysates, serum, and tissue of human origin.
Delivery information: Kit includes microtiter plate, 5X extraction reagent, standard, sample diluent, wash buffer concentrate, antibody, antibody diluent, HRP conjugate, conjugate diluent, TMB substrate, stop solution 2.