G418 disulphate

Supplier: ENZO LIFE SCIENCES
Warning

Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

ALX380013G001 ALX380013G005 ALX380013M500
ENZOALX380013G001EA 69.3 EUR
ENZOALX380013G001 ENZOALX380013G005 ENZOALX380013M500
G418 disulphate
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR