Nourseothricin sulphate (from Streptomyces noursei) ≥85%

Supplier: ENZO LIFE SCIENCES

Synonyms: NTC, Streptothricin sulphate, clonNAT

ALX380025M005
ENZOALX380025M005EA 99 EUR
ENZOALX380025M005
Nourseothricin sulphate (from Streptomyces noursei) ≥85%
Nourseothricin sulphate

Antibiotic


Active against both Gram-positive and Gram-negative bacteria and some fungi.


White to off-white powder
Mixture of streptothricin F (n=1) and D (n=3).


Soluble in water


Packaging: Shipping temperature: Ambient

Formula: C₃₁H₅₈N₁₂O₁₀·C₁₉H₃₄N₈O₈·H₂SO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00165013
CAS Number: 96736-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR