Adenosine-3’,5’-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (by HPLC)

Supplier: ENZO LIFE SCIENCES

ALX480085M001
ENZOALX480085M001EA 238 EUR
ENZOALX480085M001
Adenosine-3’,5’-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (by HPLC)
Adenosine-3’,5’-cyclic Monophosphothioate, Rp-Isomer sodium salt

Competitive inhibitor of cyclic AMP-dependent protein kinase I and II.Metabolic stability towards mammalian cyclic nucleotide- responsive phosphodiesterases.
Discriminates between protein kinase A (antagonist) and some other cyclic AMP receptors, e.g. channels or CAP 3 (agonist).Membrane-permeant for several systems (for improved permeability more lipophilic analogues e.g. Rp-8-Br-cAMPS are recommended).


  • PKA inhibitor


White to off-white powder. Please keep in mind that equal amounts of the compound may look different in volume depending on humidity. Rp-cAMPS is hygroscopic and tends to form a transparent film on the bottom of the tube.


Soluble in water or aqueous buffers, DMSO, Methanol.

Formula: C₁₀H₁₁N₅O₅PS·Na
Storage Temperature: Freezer
CAS Number: 73208-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR