Lipopolysaccharide (from Salmonella minnesota), TLRgrade™

Supplier: ENZO LIFE SCIENCES
Danger

Synonyms: Lipopolysaccharide

ALX581015L002
ENZOALX581015L002EA 313 EUR
ENZOALX581015L002
Lipopolysaccharide (from Salmonella minnesota), TLRgrade™
Lipopolysaccharides

TLR4 activator.


Rough (R)-form LPS, isolated and purified from Salmonella minnesota R345 (Rb mutant) by a modification of the PCP extraction method, converted to the uniform sodium salt form.


Strong activator of Toll-like receptor 4 (TLR4). Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice. No further re-extraction required. Smooth (S)-form LPS are commonly the preferred choice for whole animal studies, whereas Rough (R)-form LPS are primarily used in cellular in vitro activation studies.

Order Now

Specification Test Results

Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity Absence of detectable protein or DNA contaminants with agonistic TLR activity

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR