Anti-APRIL Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ALX804844C100
ENZOALX804844C100EA 500 EUR
ENZOALX804844C100
Anti-APRIL Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry

Recognizes human APRIL.

Type: Primary
Antigen: APRIL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR