Anti-UNC5A Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ALX804864C100
ENZOALX804864C100EA 534 EUR
ENZOALX804864C100
Anti-UNC5A Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry

Recognizes human UNC5A.

Type: Primary
Antigen: UNC5A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR