Caspase-2 fluorometric assay kit
Supplier: ENZO LIFE SCIENCES
ALX850214KI01
ENZOALX850214KI01EA 304 EUR
ENZOALX850214KI01
Caspase-2 fluorometric assay kit
Assays Enzyme Assays
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate VDVAD-AFC.
Delivery information: Kit includes Cell Lysis buffer (25 ml), 2X Reaction Buffer (2 ml), 1 mM VDVAD-AFC Substrate (125 µl), DTT (100 µl).