PPP ≥98% (by HPLC)

Supplier: ENZO LIFE SCIENCES

Synonyms: (5R,5aS,8aR,9R)-9-Hydroxy-5-(3,4,5-trimethoxyphenyl)-5,8,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(5aH)-one, Picropodophyllin

BMLEI3720001
ENZOBMLEI3720001EA 231 EUR
ENZOBMLEI3720001
PPP ≥98% (by HPLC)
PPP

A potent and selective inhibitor of insulin-like growth factor 1 receptor (IGF-1R, IC50=6 nM). It efficiently blocks IGF-1R activity, reduces pAKT and pERK1/2, induces apoptosis in IGF-1R-positive tumor cells and causes complete tumor regression in xenografted and allografted mice. Picropodophyllin downregulates IGF-1R by interfering with the action of β-arrestin 1/MDM2. It causes tumor regression and attenuates invasiveness of uveal melanoma cells. It displays beneficial effects on tumor growth, angiogenesis, bone disease and survival in a mouse multiple myeloma model. It attenuates intimal hyperplasia after vascular injury. Inhibition of IGF-1R is non-competitive with ATP.


  • Insulin receptor inhibitor


White solid


Soluble in DMSO (10mg/ml, warm) or dimethyl formamide (25mg/ml)

Formula: C₂₂H₂₂O₈
Storage Temperature: Freezer
MDL Number: MFCD01742647
CAS Number: 477-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84b886ff73ddde769c2ce6938fd1c6027023841d&vid=2954ca93ad41f7341fc3cb03b520210a8b0246fe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR