Doxorubicin hydrochloride ≥95% (by TLC)

Supplier: ENZO LIFE SCIENCES

BMLGR3190005 BMLGR3190025
ENZOBMLGR3190005EA 114 EUR
ENZOBMLGR3190005 ENZOBMLGR3190025
Doxorubicin hydrochloride ≥95% (by TLC)
Doxorubicin hydrochloride

Topoisomerase inhibitor


A broad spectrum antitumor agent. Induces DNA damage by intercalating into DNA and inhibiting topoisomerase II. Binds covalently to DNA. Inhbibits reverse transcriptase, RNA polymerase and the catalytic activity of Dnmt1. Immunosuppressive. Antineoplastic. Induces apoptosis. Down regulates c-myc and c-jun in doxorubicin-sensitive cells.

Formula: C₂₇H₂₉NO₁₁·HCl
MDL Number: MFCD00077757
CAS Number: 25316-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7916d2d528bf628df603af6e14cf57fb86a67e78&vid=f3e5af86cc7c215588dd5f44ace2a6e39eed57ad&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR