Cypermethrin ≥96%

Supplier: ENZO LIFE SCIENCES

BMLPR1000050
ENZOBMLPR1000050EA 303 EUR
ENZOBMLPR1000050
Cypermethrin ≥96%
Cypermethrin

Calcineurin inhibitor


Synthetic type II pyrethroid. Potent calcineurin (protein phosphatase 2B) inhibitor IC50~0.03 nM. May also have varied effects on ion channels. Serious doubts have been raised about the efficacy of this inhibitor.

Formula: C₂₂H₁₉Cl₂NO₃
MDL Number: MFCD00055328
CAS Number: 52315-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3f26e7ec246851ad28bad4acd9140bfbe0d82b8&vid=c2d8130f1c44b699406bd58737308eca95302bc3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR