epT.I.P.S.® Box 2.0 Pipette Tips
Supplier: EPPENDORF
0030076.125 0030076.133 0030076.141 0030076.150 0030076.168 0030076.176 0030076.184 0030076.192 0030076.206 0030076.214 0030076.249 0030076.257 0030076.265
EPPE0030076.125EA 42.5 EUR
EPPE0030076.125 EPPE0030076.133 EPPE0030076.141 EPPE0030076.150 EPPE0030076.168 EPPE0030076.176 EPPE0030076.184 EPPE0030076.192 EPPE0030076.206 EPPE0030076.214 EPPE0030076.249 EPPE0030076.257 EPPE0030076.265
epT.I.P.S.® Box 2.0 Pipette Tips
Pipette Tips
Eppendorf epT.I.P.S.® Box 2.0, available with 'Eppendorf Quality' pipette tips or as an empty box, improves lab efficiency with innovative TwinLid® technology, enhanced stackability, and superior ergonomics for safe and organised pipetting.
  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Boxes and trays are autoclavable at 121 °C for subsequent use
  • epT.I.P.S.® Box 2.0 and epT.I.P.S.®