Mastercycler® X50 PCR Thermocyclers
Supplier: EPPENDORF
6313000.018 6316000.019 6301000.012 6301000.047 6303000.010 6303000.044 6305000.017 6305000.041 6306000.010 6306000.045 6311000.010 6311000.045 6313000.042 6315000.040 6316000.043
EPPE6313000.018EA 9958 EUR
EPPE6313000.018 732-4394 EPPE6301000.012 EPPE6301000.047 EPPE6303000.010 EPPE6303000.044 EPPE6305000.017 EPPE6305000.041 EPPE6306000.010 EPPE6306000.045 732-3800 EPPE6311000.045 EPPE6313000.042 EPPE6315000.040 EPPE6316000.043
Mastercycler® X50 PCR Thermocyclers
PCR Cyclers
With ramp rates of up to 10 °C/s and an intuitive touchscreen, the Eppendorf® Mastercycler X50 family combines speed and flexibility, making it ideal for both routine and advanced molecular biology applications. Each cycler can be networked with nine additional eco units with independent control.
  • Heating rate: up to max. 10 °C/s
  • Wide selection of blocks from a fast silver block to 384 well block
  • VisioNize® touchscreen interface
  • Innovative 2D-Gradient for advanced PCR optimisation
  • X50 master units can control up to nine other units (connected cyclers do not