Ductless filtering chemical storage cabinets, Captair® Smart
Supplier: ERLAB CAPTAIRE
AB46A0000322 AB46A0000422 AB56A000030322 AB56A000040422
ERLAAB46A0000322EA 3589 EUR
ERLAAB46A0000322 ERLAAB46A0000422 ERLAAB56A000030322 ERLAAB56A000040422
Ductless filtering chemical storage cabinets, Captair® Smart
Fume Hoods and Enclosures Ductless Fume Hoods
The Captair® Smart filtering chemical storage cabinets are designed with a simple and innovative way of communication. Smart Technology uses simple light to show that the unit is operating safely so the user can focus his attention on what is most important: his work. Smart Technology enables visual light to indicate the hood status. A simple light pattern indicates if the air flow is compromised, if there is a fan failure, a filter breakthrough or if the doors are open.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://uk.cmd2.More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Treated air volume 200 230