Safemate ECO+ 0.9 with Perforated sectors w/s (EN12469 certified), includes UV installed on backwall, includes fixed support stand 1 * 1 items
Supplier: BioAir S.p.A.
Safemate ECO+ 0.9 with Perforated sectors w/s (EN12469 certified), includes UV installed on backwall, includes fixed support stand 1 * 1 items