Ion selective meter, handheld, Eutech Ion 6+
Supplier: EUTECH
ECION602PLUSK ECION601PLUS
662-2288EA 608 EUR
662-2288 EUTEECION601PLUS
Ion selective meter, handheld, Eutech Ion 6+
Electrochemistry Meters Ion Meters
The portable Eutech Ion 6+ offers measurements for pH, ORP, temperature and ion concentration. With its compact and sturdy design, the lightweight meter is the ideal choice for field and lab work.
  • Convenient and easy to use with pre-programmed Ion calibration points
  • mV mode allows users to check electrode calibration and performance
  • Offers a wide measurement range of up to ±500 mV
  • Compatible with a large variety of ISE electrodes
  • Shock-resistant ABS plastic casing, protected by an IP54-rated housing against accidental splashes
  • HOLD function, automatic