Anti-SPEN / SHARP Goat Polyclonal Antibody

Supplier: Everest Biotech
EB07168
EVBIEB07168EA 202 EUR
EVBIEB07168
Anti-SPEN / SHARP Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPEN / SHARP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR