Anti-lava lamp Goat Polyclonal Antibody

Supplier: Everest Biotech
EB11446
EVBIEB11446EA 339 EUR
EVBIEB11446
Anti-lava lamp Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: lava lamp
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR