Foot Sprays, Stokoderm® Foot Care
Supplier: Deb-STOKO
32780_U
EVON32780_UEA 0 EUR
EVON32780_U
Foot Sprays, Stokoderm® Foot Care
Skincare Skin Creams Hand and Body Lotions
This footspray helps to protect feet by preventing excessive perspiration, for instance when wearing closed in footwear, such as safety shoes, rubber boots etc. The alcoholic solution contains fragrances of essential oils and Hamamelis Extract, which toughens the horny layer of the feet.
  • Alcohol base provides Refresh™ing effect and helps provide effective protection against fungal skin infections
  • Essential oils provide protection against any unpleasant smells that may arise from the bacterial breakdown of sweat
  • Convenient spray form: Easy to use and apply