COLUMN GROUP 1500 ML WITH 2 OLIVES Group column 1500ml 80mm/2.5mm dia GL45 above and 2 olives 11mm below, Total height approx. 500mm, body length approx. 410mm incl. Graduation 1500/50ml, without lid 1 * 1 items
Supplier: FISCHER LABORTECHNIK
COLUMN GROUP 1500 ML WITH 2 OLIVES Group column 1500ml 80mm/2.5mm dia GL45 above and 2 olives 11mm below, Total height approx. 500mm, body length approx. 410mm incl. Graduation 1500/50ml, without lid 1 * 1 items