crystal clear, 300 strips per case Made of a special polymer with improved optical properties leading to high transmission rates ideally suited for small samples with low signal intensity Reduced shrinking during heating and cooling phases very tig 1 * 300 ST
Supplier: 4titude Ltd.
crystal clear, 300 strips per case Made of a special polymer with improved optical properties leading to high transmission rates ideally suited for small samples with low signal intensity Reduced shrinking during heating and cooling phases very tig 1 * 300 ST