AAV1/2-CMV/CBA-Null/Empty AAV Virus Vector
Supplier: Charles River Laboratories Cell Solutions
GD1004-RV
GD1004-RVEA 472 EUR
GD1004-RV
AAV1/2-CMV/CBA-Null/Empty AAV Virus Vector
Vectors, Plasmids and Libraries
AAV1/2 Vector with CMV promoter, Null/Empty, WPRE, bovine growth hormone (BGH), and polyadenylation sequence.
  • CMV Promoter
  • AAV1/2 Serotype
  • PBS with 1 mM MgCl₂ and 0.001% Lutrol F68 storage buffer
The CMV/CBA promoter consists of the chicken beta-actin promoter hybridised with the CMV immediate early enhancer sequence and is highly efficient in most tissue types. The Woodchuck post-transcriptional
regulatory element (WPRE) and the presence of a bovine growth hormone (BGH) polyadenylation sequence ensures high transcription following transduction.
Shipped in dry ice.