Butyrometer Waterbath

Supplier: Gerber Instruments AG
03.9202
GEAG03.9202EA 1800 EUR
GEAG03.9202
Butyrometer Waterbath
Baths Water Baths, Unstirred
Waterbath for tempering liquids with continuous microprocessor control for easy operation. Radiator made of stainless steel , incl. ground grid, 7-segment LED display.

  • Material: Made of white PP, thickness 10 mm
  • Temperature range: 10 °C above ambient to 95 °C (±0,3)
  • Power: 1100 W
  • Voltage: 230 V/50 Hz

Display actual value, set point value via one keypress, input set point value in 0,1 °C increments. Shutdown in case of overheating, sensor damage or lack of water.

Available with circulation pump (option on request) to improve the temperature distribution.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR