An ELISA kit for the detection of SPA (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 9.375pg/ml. 1 * 1 KIT
Supplier: G Bioscienes
An ELISA kit for the detection of SPA (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 9.375pg/ml. 1 * 1 KIT