Anti-B-tag Rabbit Polyclonal Antibody

Supplier: Genscript
A00638-40
GENSA00638-40EA 173 EUR
GENSA00638-40
Anti-B-tag Rabbit Polyclonal Antibody
Antibodies
Type: Tag
Antigen: B-tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR