Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody (Biotin)

Supplier: Genscript
A01737-100
GENSA01737-100EA 598 EUR
GENSA01737-100
Anti-NWSHPQFEK Tag Mouse Monoclonal Antibody (Biotin)
Antibodies
Type: Tag
Antigen: NWSHPQFEK Tag
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR