Straw pipettes, Corning® Gosselin™
Supplier: Corning
EPI24B-01 EPI5-01 EPI1B-05 EPI24B-02 EPI1B-03
GOSSEPI24B-01EA 641 EUR
GOSSEPI24B-01 GOSSEPI5-01 GOSSEPI1B-05 GOSSEPI24B-02 GOSSEPI1B-03
Straw pipettes, Corning® Gosselin™
Pipettes Straw Pipettes
Straw pipettes made of PP for liquid handling.
  • Packed in tamper-evident zip bags
  • Sterile (SAL 10-3)
  • Universal size
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904