Swabs in tubes
Supplier: Greiner Bio-One
421161_1000 420161_1300 425010_1000
391-3795EA 409 EUR
391-3795 GREI420161_1300 GREI425010_1000
Swabs in tubes
Swabs Swabs in Tubes
These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
  • Sterilised by irradiation
  • Tubes are made of polystyrene
  • Swabs are made of cotton
Suitable for hygienic controls in food industry as well as in environmental sampling and veterinary research.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 (241002); ISO 50001 (241002)