1-Phenylethyl mercaptan 98%

Supplier: Thermo Fisher Scientific
Danger

H26345.03 H26345.09
H26345.03EA 46 EUR
H26345.03 H26345.09
1-Phenylethyl mercaptan 98%
1-Phenylethyl mercaptan

Purity: 98% CAS No.: 6263-65-6 Molecular Formula: C8H10S Formula Weight: 138.23 Flash Point: 76°(169°F) Refractive Index: 1.5580 UN No.: UN2810 MDL No.: MFCD00053515

Formula: C₈H₁₀S
Density: 1.021
Flash Pt: 76 °C (169 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00053515
CAS Number: 6263-65-6
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR