5-Bromo-2'-deoxyuridine (broxuridine) ≥99%

Supplier: Thermo Fisher Scientific
Warning

H27260.03 H27260.06 H27260.MD
H27260.03EA 124 EUR
H27260.03 H27260.06 H27260.MD
5-Bromo-2'-deoxyuridine (broxuridine) ≥99%
5-Bromo-2'-deoxyuridine (Broxuridine)

Purity: 99% CAS No.: 59-14-3 Molecular Formula: C9H11BrN2O5 Formula Weight: 307.10 Storage Temperature: 2°C to 8°C Melting Point: ca 190° dec. Beilstein Reference: 30395 Merck Reference: 14,1452 Optical Rotation: +31° (c=1 in 0.1N HCl) MDL No.: MFCD00006529

Formula: C₉H₁₁BrN₂O₅
Melting Pt: ∼ 190 °C (dec.)
MDL Number: MFCD00006529
CAS Number: 59-14-3
EINECS: 200-415-9
Merck Index: 14,1452

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR