4-(Pyrrolidin-3-yl)morpholine 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-(3-Pyrrolidinyl)morpholine

H51002.03 H51002.MD
H51002.03EA 239 EUR
H51002.03 H51002.MD
4-(Pyrrolidin-3-yl)morpholine 97%
4-(Pyrrolidin-3-yl)morpholine

Purity: 97% CAS No.: 53617-37-1 Molecular Formula: C8H16N2O Formula Weight: 156.23 UN No.: UN2735 MDL No.: MFCD06740352

Formula: C₈H₁₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD06740352
CAS Number: 53617-37-1
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR