1-Methylsulphonyl-1H-benzotriazole 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-(Methylsulphonyl)-1H-benzo[d][1,2,3]triazole

H53386.03 H53386.06 H53386.14
H53386.03EA 42.5 EUR
H53386.03 H53386.06 H53386.14
1-Methylsulphonyl-1H-benzotriazole 97%
1-Methylsulphonyl-1H-benzotriazole

Purity: 97% CAS No.: 37073-15-7 Molecular Formula: C7H7N3O2S Formula Weight: 197.21 Melting Point: 106-108° dec. MDL No.: MFCD00595638

Formula: C₇H₇N₃O₂S
Melting Pt: 106…108 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00595638
CAS Number: 37073-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR