Chlorotris(triphenylphosphine)cobalt(I) 97%

Supplier: Thermo Fisher Scientific
Warning

H55395.06
H55395.06EA 177 EUR
H55395.06
Chlorotris(triphenylphosphine)cobalt(I) 97%
Chlorotris(triphenylphosphine)cobalt(I)

Purity: 97% CAS No.: 26305-75-9 Molecular Formula: C54H45ClCoP3 Formula Weight: 881.26 Melting Point: 135-139° MDL No.: MFCD00015864

Formula: C₅₄H₄₅ClCoP₃
MW: 881.26 g/mol
Melting Pt: 135…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00015864
CAS Number: 26305-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR