(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 98%

Supplier: Thermo Fisher Scientific

Synonyms: Boc-Tyr(Me)-OH, N-Boc-O-methyl-L-tyrosine

H61820.03 H61820.06
H61820.03EA 68.6 EUR
H61820.03 H61820.06
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid

Purity: 98% CAS No.: 53267-93-9 Molecular Formula: C15H21NO5 Formula Weight: 295.33 Physical Form: Solid MDL No.: MFCD00065603

Formula: C₁₅H₂₁NO₅
Storage Temperature: Ambient
MDL Number: MFCD00065603
CAS Number: 53267-93-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR