4-Chlorophenethylamine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(4-Chlorophenyl)ethylamine

H63081.06 H63081.14 H63081.22
H63081.06EA 31.1 EUR
H63081.06 H63081.14 H63081.22
4-Chlorophenethylamine 97%
4-Chlorophenethylamine
Formula: C₈H₁₀ClN
Boiling Pt: 60…65 °C (0.1 mmHg)
Density: 1.120
Flash Pt: 106 °C (223 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008191
CAS Number: 156-41-2
EINECS: 205-853-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR