Methyl-6-chloropicolinate 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 6-chloropyridine-2-carboxylate, Methyl 6-chloropicolinate

H64023.06 H64023.14
H64023.06EA 53.6 EUR
H64023.06 H64023.14
Methyl-6-chloropicolinate 95%
Methyl 6-chloropicolinate

Purity: 95% CAS No.: 6636-55-1 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Pale yellow Melting Point: 92-98° lit. Beilstein Reference: 128580 MDL No.: MFCD00155391

Formula: C₇H₆ClNO₂
Melting Pt: 92…98 °C (lit.)
MDL Number: MFCD00155391
CAS Number: 6636-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR