Sodium thiosulphate 0.2 mol/l (0.2 N) in aqueous solution

Supplier: Hach

Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate

2267501
HACH2267501EA 38.3 EUR
HACH2267501
Sodium thiosulphate 0.2 mol/l (0.2 N) in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR