Visual comparator colour kits, phenols
Supplier: Hach
2483600
HACH2483600EA 298 EUR
HACH2483600
Visual comparator colour kits, phenols
Environmental Test Kits
Simple, economical, rapid visual colour determination.
Delivery information: Kit includes four reagents, two tubes, two color discs, color comparator box, instruction sheet, and carrying case.