Sulphuric acid 0.015 mol/l (0.03 N) in aqueous solution

Supplier: Hach

2620532 2620553
HACH2620532EA 23.6 EUR
HACH2620532 HACH2620553
Sulphuric acid 0.015 mol/l (0.03 N) in aqueous solution
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR