Bi-metallic dial thermometer
Supplier: Hach
4565500
HACH4565500EA 116 EUR
HACH4565500
Bi-metallic dial thermometer
Thermometers Analogue Thermometers
Bi-metallic dial gauge 0 to 200 °C ±1%. Two degree divisions.
Probe Ø 381 mm.
Packaging: Plastic carry case with clip.