Ion selective electrode, ammonia (NH₃), gas-sensing, IntelliCAL™
Supplier: Hach
ISENH318101 ISENH3181AP ISENH318103
HACHISENH318101EA 973 EUR
HACHISENH318101 HACHISENH3181AP HACHISENH318103
Ion selective electrode, ammonia (NH₃), gas-sensing, IntelliCAL™
Electrodes ISE Electrodes
This digital, combination gas-sensing ammonia ion selective electrode (ISE) with replaceable membrane module, refillable outer body, double junction reference and built-in temperature sensor is intended for laboratory use.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Refillable outer body, easy to maintain modular membrane design, and large annular reference junction for maximum surface area between sample and reference electrolyte provide great measurement stability
  • Ultimate traceability in measurement history: Stores time, date stamp,