Shaking water baths, Precision™
Supplier: THERMO ELECTRON LED
TSSWB15 TSSWB27 TSSWB15S TSDUB15
HAKATSSWB15EA 2550 EUR
HAKATSSWB15 HAKATSSWB27 HAKATSSWB15S HAKATSDUB15
Shaking water baths, Precision™
Baths Shaking Water Baths
Precision™ water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing.
  • Coil-free interior for easy cleaning
  • Four temperature and shaking speed presets
  • Audible alarms protect work
  • Auto-on and auto-off detectors
  • Low-fluid detection
  • Monitor with icon based graphic display
The shallow form shaking bath has a removable tray that allows it to be used with smaller sample containers. Dubnoff shaking water baths are designed specifically for applications that requireMore Product Information