Sieve Shakers, Heavy Duty, UWL
Supplier: NEXOPART
510-0969EA 7580 EUR
510-0969 510-0972 510-0966 510-0971 510-0974 510-0968 510-1305 510-1304 510-1302 510-1307 HAVE205323102 HAVE205323133 HAVE205323140 HAVE205322846 HAVE205322853
Sieve Shakers, Heavy Duty, UWL
Shakers Sieve Shakers
For test sieves up to 400 mm diameter or square sieves up to 500×500 mm. The UWL 400 is an efficient test sieve shaker that achieves optimum results for large feed quantities. It creates a three-dimensional sieving action, and is especially designed for coarse grained products. The UWL 400 T is suitable for dry sieving; the UWL 400 N is suitable for wet sieving, and the UWL 400 H is suitable for square test sieves with beechwood frames.
  • Separate control device, wall fixing possible
  • Rotation speed: 1500 min⁻¹
  • Timer function: 0 to 599 minutes
  • Max.