Reagent reservoirs
Supplier: HEATHROW
120753 120750 120752
HEAT120753EA 86.2 EUR
HEAT120753 HEAT120750 HEAT120752
Reagent reservoirs
Reagent Reservoirs
These disposable reagent reservoirs have a fresh and modern design with enhanced features to streamline pipetting procedures.
  • Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sterile,