Blood sedimentation pipettes, acc. to Westergren-Katz
Supplier: GLASWARENFABRIK KARL HECHT
40380015 40380010
612-0593EA 1.7 EUR
612-0593 HECH40380010
Blood sedimentation pipettes, acc. to Westergren-Katz
Pipettes Haemometer Pipettes
Blood sedimentation pipettes for the macro method.
  • Clear glass
  • Fused blue graduation 'ASSISTENT-PERMANENT'
  • Division 0 to 200 mm: 1 mm
Certifications: DIN EN ISO 13079