Thermostatic water bath
Supplier: GLASWARENFABRIK KARL HECHT
43180010
HECH43180010EA 1490 EUR
HECH43180010
Thermostatic water bath
Baths Boiling Baths
The Thermostatic waterbath, CE certified is fully electronic wear-free temperature control with digital display. Temperature control with adjustable nominal value limit and alarm circuit.
  • Themperature stability of ± 0,1 K
  • Operating temperature range with external cooling of −30 °C upto +100 °C