Attachment or rack for holding 20 test tubes with a diameter of 17 - 20 mm. Material: electro-polished stainless steel Characteristics: autoclavable and dishwasher safe angle adjustment up to 90° attachment without foot part can be used as table fr 1 * 1 items
Supplier: Heidolph Instruments GmbH & Co.KG
Attachment or rack for holding 20 test tubes with a diameter of 17 - 20 mm. Material: electro-polished stainless steel Characteristics: autoclavable and dishwasher safe angle adjustment up to 90° attachment without foot part can be used as table fr 1 * 1 items
New Product