Thermo Scientific™ Maxisafe 2030i Biological Safety Cabinets
Supplier: Thermo Scientific
51032715 51032716 51032717 51032718 51032711 51032712 51032713 51032714
HERA51032715EA 27360 EUR
HERA51032715 HERA51032716 HERA51032717 HERA51032718 135-2908 135-2909 135-2910 135-2911
Thermo Scientific™ Maxisafe 2030i Biological Safety Cabinets
Fume Hoods and Enclosures Biological Safety Cabinets
The Maxisafe 2030i incorporates SmartFlow Plus, DAVe Plus and Smart UV technologies for ultimate protection and graphical user interface for ease of operation. Each unit has inbuilt Wifi capability and is ready to be connected. Our prefilter system includes a new improved and easy sealing solution.
  • 7" GUI touchscreen interface
  • LED interior illumination
  • Cloud ready data output
  • Ergonomic arm rests (option)
  • SmartPort versatile access
  • SmartFlow plus auto compensation
  • 316 stainless steel interior
  • Previous models: Herasafe KSP
Certifications: EN 12469, DIN12980