Capillary tubings, stainless steel, J&W UltiMetal plus
Supplier: VARIAN
CP6577 CP6580 CP6575 CP6572 CP6578 CP6579 CP6576
VARICP6577EA 177 EUR
VARICP6577 VARICP6580 VARICP6575 HEWLCP6572 VARICP6578 VARICP6579 VARICP6576
Capillary tubings, stainless steel, J&W UltiMetal plus
Tubing Chromatography Tubing
UltiMetal Plus Stainless Steel Capillary Tubing products are the preferred choice for most applications due to its inertness and robustness.
  • UltiMetal Plus deactivation
  • Chemical deactivation provides an inert surface for analysis of active analytes at trace levels
  • Virtually unbreakable stainless steel tubing which provides robustness
  • Three types of products available: transfer lines, with ¹/₁₆" outer diameter, guard columns and 50 m capillary tubing
These tubing products can be used as transfer lines, guard columns or for making connections in GC. TheMore Product Information