Extended light path (bubble cell) bare fused-silica capillaries
Supplier: Agilent
G1600-60132 G1600-60232 G1600-60332 G1600-61132 G1600-61232 G1600-61332 G1600-62132 G1600-62232 G1600-62332 G1600-64232 G1600-64332 G1600-60233
HEWLG1600-60132EA 205 EUR
HEWLG1600-60132 HEWLG1600-60232 HEWLG1600-60332 HEWLG1600-61132 HEWLG1600-61232 HEWLG1600-61332 HEWLG1600-62132 HEWLG1600-62232 HEWLG1600-62332 HEWLG1600-64232 HEWLG1600-64332 HEWLG1600-60233
Extended light path (bubble cell) bare fused-silica capillaries
Electrophoresis Systems Capillary Electrophoresis Systems
Agilent Technologies extended light path capillaries ('bubble' cell capillaries) can be used to improve sensitivity 3- to 5-fold over standard capillaries. With extended light path capillaries, the inner diameter is increased only at the detection window, offering the sensitivity of a wide inner diameter capillary and the low current generation of a narrow one.
Resolution is not sacrificed when used with matching optical alignment interfaces from Agilent.
Through a computer-controlled proprietary process, the diameter is increased three to five times, with a manufacturing precisionMore Product Information